missing translation for 'onlineSavingsMsg'
Learn More

(4S,5R)-3-(tert-Butoxycarbonyl)-2,2-dimethyl-4-phenyl-1,3-oxazolidine-5-carboxylic Acid 98.0+%, TCI America™

Catalog No. b4127200mg
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4127200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B4127200MG Supplier TCI America Supplier No. B4127200MG

Chemical Identifiers

CAS 143527-70-2
Molecular Formula C17H23NO5
Molecular Weight (g/mol) 321.373
MDL Number MFCD09263630
InChI Key VAHXMEZCPGHDBJ-QWHCGFSZSA-N
Synonym (4S,5R)-3-Boc-2,2-dimethyl-4-phenyl-1,3-oxazolidine-5-carboxylic Acid
PubChem CID 10969286
IUPAC Name (4S,5R)-2,2-dimethyl-3-[(2-methylpropan-2-yl)oxycarbonyl]-4-phenyl-1,3-oxazolidine-5-carboxylic acid
SMILES CC1(N(C(C(O1)C(=O)O)C2=CC=CC=C2)C(=O)OC(C)(C)C)C

Specifications

Melting Point 114°C
Color White-Yellow
Quantity 200 mg
Formula Weight 321.37
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (4S,5R)-3-(tert-Butoxycarbonyl)-2,2-dimethyl-4-phenyl-1,3-oxazolidine-5-carboxylic Acid
TSCA No
Recommended Storage Refrigerator