accessibility menu, dialog, popup

UserName

CAS RN 143527-70-2

IUPAC Name: 3-[(tert-butoxy)carbonyl]-2,2-dimethyl-4-phenyl-1,3-oxazolidine-5-carboxylic acid
Synonyms: 3-(tert-butoxycarbonyl)-2,2-dimethyl-4-phenyl-1,3-oxazolidine-5-carboxylic acid
Molecular Weight (g/mol): 321.37
Molecular Formula: C17H23NO5
InChi Key: VAHXMEZCPGHDBJ-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1C(C(OC1(C)C)C(O)=O)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results