missing translation for 'onlineSavingsMsg'
Learn More

(4S,4'S)-2,2'-(Pentane-3,3'-diyl)bis(4-benzyl-4,5-dihydrooxazole) 98.0+%, TCI America™

Supplier:  TCI America P19471G

 View more versions of this product

Catalog No. P19471G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

(4S,4′S)-2,2′-(Pentane-3,3′-diyl)bis(4-benzyl-4,5-dihydrooxazole)
Yellow
Clear Liquid at 20°C
(S,S)-3,3-Bis(4-benzyl-2-oxazolin-2-yl)pentane, (S,S)-2,2′C-(Diethylmethylene)bis(4-benzyl-2-oxazoline)
CCC(CC)(C1=NC(CO1)CC2=CC=CC=C2)C3=NC(CO3)CC4=CC=CC=C4
390.527
390.53
160191-64-0
1 g
C25H30N2O2
ZIDQHNROIGAKIB-VXKWHMMOSA-N
(4S)-4-benzyl-2-[3-[(4S)-4-benzyl-4,5-dihydro-1,3-oxazol-2-yl]pentan-3-yl]-4,5-dihydro-1,3-oxazole
10960121
≥98.0% (T)

Chemical Identifiers

160191-64-0
390.527
(S,S)-3,3-Bis(4-benzyl-2-oxazolin-2-yl)pentane, (S,S)-2,2′C-(Diethylmethylene)bis(4-benzyl-2-oxazoline)
(4S)-4-benzyl-2-[3-[(4S)-4-benzyl-4,5-dihydro-1,3-oxazol-2-yl]pentan-3-yl]-4,5-dihydro-1,3-oxazole
C25H30N2O2
ZIDQHNROIGAKIB-VXKWHMMOSA-N
10960121
CCC(CC)(C1=NC(CO1)CC2=CC=CC=C2)C3=NC(CO3)CC4=CC=CC=C4
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions