accessibility menu, dialog, popup

UserName

CAS RN 160191-64-0

IUPAC Name: 4-benzyl-2-[3-(4-benzyl-4,5-dihydro-1,3-oxazol-2-yl)pentan-3-yl]-4,5-dihydro-1,3-oxazole
Synonyms: 4-benzyl-2-[3-(4-benzyl-4,5-dihydro-1,3-oxazol-2-yl)pentan-3-yl]-4,5-dihydro-1,3-oxazole
Molecular Weight (g/mol): 390.53
Molecular Formula: C25H30N2O2
InChi Key: ZIDQHNROIGAKIB-UHFFFAOYNA-N
SMILES: CCC(CC)(C1=NC(CC2=CC=CC=C2)CO1)C1=NC(CC2=CC=CC=C2)CO1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)