missing translation for 'onlineSavingsMsg'
Learn More

(4S,4'S)-2,2'-(Pentane-3,3'-diyl)bis(4-benzyl-4,5-dihydrooxazole) 98.0+%, TCI America™

Catalog No. P1947200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1947200MG 200 mg
P19471G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P1947200MG Supplier TCI America Supplier No. P1947200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 160191-64-0
Molecular Formula C25H30N2O2
Molecular Weight (g/mol) 390.527
InChI Key ZIDQHNROIGAKIB-VXKWHMMOSA-N
Synonym (S,S)-3,3-Bis(4-benzyl-2-oxazolin-2-yl)pentane, (S,S)-2,2′C-(Diethylmethylene)bis(4-benzyl-2-oxazoline)
PubChem CID 10960121
IUPAC Name (4S)-4-benzyl-2-[3-[(4S)-4-benzyl-4,5-dihydro-1,3-oxazol-2-yl]pentan-3-yl]-4,5-dihydro-1,3-oxazole
SMILES CCC(CC)(C1=NC(CO1)CC2=CC=CC=C2)C3=NC(CO3)CC4=CC=CC=C4

Specifications

Color Yellow
Quantity 200 mg
Physical Form Clear Liquid at 20°C
Formula Weight 390.53
Percent Purity ≥98.0% (T)
Chemical Name or Material (4S,4′S)-2,2′-(Pentane-3,3′-diyl)bis(4-benzyl-4,5-dihydrooxazole)