missing translation for 'onlineSavingsMsg'
Learn More

(4S)-(-)-4-Isopropyl-2-oxazolidinone, 98%

Catalog No. AC297860010
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC297860010 1 g Glass Bottle
AC297860050 5 g Glass Bottle
AC297860250 25 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC297860010 Supplier Thermo Scientific Chemicals Supplier No. 297860010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 17016-83-0 | C6H11NO2 | 129.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17016-83-0
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.16
MDL Number MFCD00010847
InChI Key YBUPWRYTXGAWJX-RXMQYKEDSA-N
Synonym s-4-isopropyl-2-oxazolidinone, s---4-isopropyl-2-oxazolidinone, 4s---4-isopropyl-2-oxazolidinone, s-4-isopropyloxazolidin-2-one, 4s-4-isopropyl-1,3-oxazolidin-2-one, 2-oxazolidinone, 4-1-methylethyl-, 4s, 4s-4-propan-2-yl-1,3-oxazolidin-2-one, 4s---isopropyl-2-oxazolidinone, 4 s-1-methylethyl-2-oxazolidinone, 4s---4-isopropyl-2-oxazolidinon
PubChem CID 7157133
IUPAC Name (4S)-4-propan-2-yl-1,3-oxazolidin-2-one
SMILES CC(C)C1COC(=O)N1

Specifications

Melting Point 70°C to 73°C
CAS Min % 97.5
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Specific Rotation -18
Solubility Information Solubility in water: insoluble in water
Specific Rotation Condition −18° (20°C c=1,ethanol)
Quantity 1 g
Formula Weight 129.16
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material (4S)-(-)-4-Isopropyl-2-oxazolidinone
Show More Show Less

RUO – Research Use Only