missing translation for 'onlineSavingsMsg'
Learn More

(4R)-(+)-4-Isopropyl-2-oxazolidinone, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 297850050

 View more versions of this product

Catalog No. AC297850050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

(4R)-(+)-4-Isopropyl-2-oxazolidinone
97.5
Authentic
Glass bottle
MFCD00075172
−18° (20°C c=1,ethanol)
r-4-isopropyl-2-oxazolidinone, r-4-isopropyloxazolidin-2-one, r-+-4-isopropyl-2-oxazolidinone, 4r-+-4-isopropyl-2-oxazolidinone, 4r-4-isopropyl-1,3-oxazolidin-2-one, 2-oxazolidinone, 4-1-methylethyl-, 4r, 4r-4-propan-2-yl-1,3-oxazolidin-2-one, pubchem8176, r-isopropyloxazolidinone
YBUPWRYTXGAWJX-YFKPBYRVSA-N
(4R)-4-propan-2-yl-1,3-oxazolidin-2-one
641505
98%
95530-58-8
100.0
97.5% min. (GC)
C6H11NO2
5g
-18
Solubility in water: soluble in water
CC(C)[C@@H]1COC(=O)N1
129.16
129.16

Chemical Identifiers

95530-58-8
129.16
YBUPWRYTXGAWJX-YFKPBYRVSA-N
641505
CC(C)[C@@H]1COC(=O)N1
C6H11NO2
MFCD00075172
r-4-isopropyl-2-oxazolidinone, r-4-isopropyloxazolidin-2-one, r-+-4-isopropyl-2-oxazolidinone, 4r-+-4-isopropyl-2-oxazolidinone, 4r-4-isopropyl-1,3-oxazolidin-2-one, 2-oxazolidinone, 4-1-methylethyl-, 4r, 4r-4-propan-2-yl-1,3-oxazolidin-2-one, pubchem8176, r-isopropyloxazolidinone
(4R)-4-propan-2-yl-1,3-oxazolidin-2-one
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only