missing translation for 'onlineSavingsMsg'
Learn More

(4R)-(+)-4-Isopropyl-2-oxazolidinone, 98%

Catalog No. AC297850010
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC297850010 1 g Glass Bottle
AC297850050 5 g Glass Bottle
AC297850250 25 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC297850010 Supplier Thermo Scientific Chemicals Supplier No. 297850010
Only null left

CAS: 95530-58-8 | C6H11NO2 | 129.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 95530-58-8
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.16
MDL Number MFCD00075172
InChI Key YBUPWRYTXGAWJX-YFKPBYRVSA-N
Synonym r-4-isopropyl-2-oxazolidinone, r-4-isopropyloxazolidin-2-one, r-+-4-isopropyl-2-oxazolidinone, 4r-+-4-isopropyl-2-oxazolidinone, 4r-4-isopropyl-1,3-oxazolidin-2-one, 2-oxazolidinone, 4-1-methylethyl-, 4r, 4r-4-propan-2-yl-1,3-oxazolidin-2-one, pubchem8176, r-isopropyloxazolidinone
PubChem CID 641505
IUPAC Name (4R)-4-propan-2-yl-1,3-oxazolidin-2-one
SMILES CC(C)[C@@H]1COC(=O)N1

Specifications

Melting Point 70°C to 73°C
CAS Min % 97.5
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Specific Rotation -18
Specific Rotation Condition −18° (20°C c=1,ethanol)
Solubility Information Solubility in water: soluble in water
Formula Weight 129.16
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material (4R)-(+)-4-Isopropyl-2-oxazolidinone
Show More Show Less

RUO – Research Use Only