missing translation for 'onlineSavingsMsg'
Learn More

(4aS,8aS)-2-Benzoyl-1,3,4,4a,5,8a-hexahydro-6(2H)-isoquinolinone 97.0+%, TCI America™

Catalog No. b2360100mg
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B2360100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B2360100MG Supplier TCI America Supplier No. B2360100MG
Only null left

Chemical Identifiers

CAS 52346-14-2
Molecular Formula C16H17NO2
Molecular Weight (g/mol) 255.317
MDL Number MFCD06797067
InChI Key KIGUODXTRRRCCD-KBPBESRZSA-N
PubChem CID 10879687
IUPAC Name (4aS,8aS)-2-benzoyl-1,3,4,4a,5,8a-hexahydroisoquinolin-6-one
SMILES C1CN(CC2C1CC(=O)C=C2)C(=O)C3=CC=CC=C3

Specifications

Quantity 100 mg
Formula Weight 255.32
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (4aS,8aS)-2-Benzoyl-1,3,4,4a,5,8a-hexahydro-6(2H)-isoquinolinone