accessibility menu, dialog, popup

UserName

CAS RN 52346-14-2

IUPAC Name: 2-benzoyl-1,2,3,4,4a,5,6,8a-octahydroisoquinolin-6-one
Synonyms: 2-benzoyl-1,3,4,4a,5,8a-hexahydroisoquinolin-6-one
Molecular Weight (g/mol): 255.32
Molecular Formula: C16H17NO2
InChi Key: KIGUODXTRRRCCD-UHFFFAOYNA-N
SMILES: O=C(N1CCC2CC(=O)C=CC2C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results