missing translation for 'onlineSavingsMsg'
Learn More

Spectrum™ Labs Spectra/Por™ 6 3500 D MWCO Standard RC Pre-wetted Dialysis Kits

Catalog No. 08700191
Change view
Click to view available options
Diameter (Metric):
11.5 mm
29 mm
34 mm
Length (Metric):
1.1 mL/cm vol./length
6.4 mL/cm vol./length
9.3 mL/cm vol./length
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Length (Metric)
08700191 34 mm 9.3 mL/cm vol./length
08700190 29 mm 6.4 mL/cm vol./length
08700189 11.5 mm 1.1 mL/cm vol./length
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 08700191 Supplier Spectrum™ Supplier No. 132594T

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

  • 1 meter of Spectra/Por 6 Dialysis Tubing
  • 1 Standard Spectra/Por Closure (for buoyancy)
  • 1 Weighted Spectra/Por Closure (for orientation)
  • 5 Tubing Opening Picks

With Pre-wetted ready-to-use tubing, not requiring soaking to remove glycerol

  • Clear, flexible, study standard grade Regenerated Cellulose (RC) dialysis membrane
  • Membrane packaged wet in preservative solution of 0.05% sodium azide
  • Kits available in 1m tubing lengths ideal for membrane evaluation and qualification
  • Include a pair of closures and convenient tubing opening picks

Specifications

Product Type Dialysis Trial Kit
MWCO 3.5 kDa
Format Tubing
Diameter (Metric) 34 mm
Length (Metric) 9.3 mL/cm vol./length
Quantity 1 m Tubing/Kit