Learn More
Fisherbrand™ Filtration Extraction Thimbles - Grade 645 Cellulose
Ensures complete extraction of samples, prevents contamination and minimize hands-on time
$190.55 - $284.97
Specifications
| Quantity | 25 |
|---|---|
| Description | Filtration Extraction Thimbles - Grade 645 Cellulose |
| Bottom Shape | Round |
| Material | Cellulose |
| For Use With (Application) | Fat determination in food and feed, Determination of binder content, Analysis of soil contaminants, Determination of additives in rubber and elastomers |
| Catalog Number | Diameter (Metric) Inside | Length (Metric) | Wall Thickness | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Diameter (Metric) Inside | Length (Metric) | Wall Thickness | Price | Quantity & Availability | ||||
|
12101106
|
33 mm | 118 mm | 1 mm |
Pack of 25 Each for $284.97
|
|
||||
|
12101110
|
43 mm | 123 mm | 2 mm |
Pack of 25 Each for $284.97
|
|
||||
|
12101105
|
43 mm | 123 mm | 1 mm |
Pack of 25 Each for $284.97
|
|
||||
|
12101103
|
33 mm | 80 mm | 1 mm |
Pack of 25 Each for $190.55
|
|
||||
|
12101101
|
25 mm | 80 mm | 1 mm |
Pack of 25 Each for $190.55
|
|
||||
|
12101100
|
22 mm | 80 mm | 1 mm |
Pack of 25 Each for $190.55
|
|
||||
|
12101102
|
26 mm | 60 mm | 1 mm |
Pack of 25 Each for $190.55
|
|
||||
|
12101104
|
33 mm | 94 mm | 1 mm |
Pack of 25 Each for $190.55
|
|
||||
Description
Ensures complete extraction of samples, prevents contamination and minimize hands-on time
- Used for Soxhlet extractors or automized extractors
- Minimizes the hands-on time
- Ensures a complete extraction
- Special selected raw materials
- High purity - low contamination
- Effectively protects the extract
- Safe and reliable extraction
- Mechanical strength and excellent retention capacities
- Perfect fit for convenient workflow
- Optimal usability for special applications
Specifications
| 25 | |
| Round | |
| Fat determination in food and feed, Determination of binder content, Analysis of soil contaminants, Determination of additives in rubber and elastomers | |
| Soxhlet extractors or automized extractors |
| Filtration Extraction Thimbles - Grade 645 Cellulose | |
| Cellulose | |
| 645 |