missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ SXT Oil Only Absorbent Pads
Made to handle petroleum-based spills
$119.11
Specifications
| Material | Polypropylene |
|---|---|
| Type | Absorbent Pad, Oil-only |
| For Use With (Application) | Overspray leaks and drips, workstations, heavy clean-up and wipe-up , drums - liquid storage, outfalls and discharge ponds, food processing, high tech industries, machining of small parts, paper industry, pharmaceutical |
| Dimensions (L x W) | 38.1 x 48.2 cm (15 x 19 in.) |
| Catalog Number | Mfr. No. | Capacity (English) | Weight Capacity | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (English) | Weight Capacity | Price | Quantity & Availability | ||||
|
17111132
|
Brady™
SXT300 |
17 gal./Bale | Medium-weight |
Case of 100 Each for $119.11
|
|
||||
Description
Specifications
| Polypropylene | |
| Absorbent Pad, Oil-only | |
| Overspray leaks and drips, workstations, heavy clean-up and wipe-up , drums - liquid storage, outfalls and discharge ponds, food processing, high tech industries, machining of small parts, paper industry, pharmaceutical | |
| 38.1 x 48.2 cm (15 x 19 in.) |