missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Oil Plus™ Oil Only Heavy Rolls
Description
Specifications
Specifications
| Design | Triple Perforated |
| Capacity (English) | 9 gal./Bale |
| Material | Polypropylene |
| Type | Oil Only Heavy Roll |
| For Use With (Application) | Overspray leaks and drips, workstations, heavy clean-up and wipe-up, drums - liquid storage, outfalls and discharge ponds |
| Packaging Quantity | 1 Ea. |
| Weight Capacity | Heavyweight |
| Dimensions (L x W) | 50 ft. x 15 in. |