Learn More
Brady™ MRO Plus™ Universal Heavy Pads
Catalog No. 17111151
Change view
Click to view available options
Capacity (English):
26 gal.
Catalog No. Capacity (English)
17111151 26 gal.
1 options
Catalog No. 17111151 Supplier Brady™ Supplier No. MRO100B
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Absorbs all industrial liquids (water, petroleum-based and chemical-based fluids)
  • Three-ply construction
  • Dimpled and Perforated - reduce waste
  • Gray color masks drips and spills
  • Specifications
    Color Gray
    Capacity (English) 26 gal.
    Material Polypropylene
    Type Universal Absorbent Pad
    For Use With (Application) Overspray leaks and drips, workstations, heavy clean-up and wipe-up, drums - liquid storage, outfalls and discharge ponds
    Packaging Quantity 100/Cs.
    Dimensions (L x W) 38 x 48.26 cm (15 x 19 in.)