missing translation for 'onlineSavingsMsg'
Learn More

Brady™ GP™ MAXX Enhanced Universal Absorbent Pads

Catalog No. 17111153
Change view
Click to view available options
Capacity (English):
25 gal.
30 gal.
44 gal.
Packaging Quantity:
100/Bag
100/Cs.
200/Bag
Weight Capacity:
Heavyweight
Lightweight
Medium-weight
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Packaging Quantity Weight Capacity
17111153 30 gal. 100/Cs. Heavyweight
17111148 44 gal. 200/Bag Lightweight
17111149 25 gal. 100/Cs. Medium-weight
17111147 30 gal. 100/Bag Heavyweight
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 17111153 Supplier Brady™ Supplier No. GP100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

General purpose absorbent for all fluids (water, petroleum and chemical-based)

  • Single-ply construction
  • Class: Universal
  • Available in various weights, (Heavy, Medium and Light)
  • Made with MAXX: greater performance without the extra weight

    Specifications

    Certifications/Compliance Pass 2011/65/EU
    Color Gray
    Capacity (English) 30 gal.
    Capacity (Metric) 30 gal./Cs.
    Material Polypropylene
    Type Absorbent Pad
    For Use With (Application) Overspray leaks and drips, workstations, drums - liquid storage, outfalls and discharge Ponds
    Packaging Quantity 100/Cs.
    Weight Capacity Heavyweight
    Dimensions (L x W) 38 x 48.26 cm (15 x 19 in.)