missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Bench Complete ESD Accessories Kit

Numéro de catalogue. 25100323
Change view
Click to view available options
Width (English):
48 in.
60 in.
72 in.
Width (Metric):
122 cm
152.4 cm
182.8 cm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Width (English) Width (Metric)
25100323 48 in. 122 cm
25100324 60 in. 152.4 cm
25100325 72 in. 182.8 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. 25100323 Fournisseur Fisherbrand™ Code fournisseur 48ESDFSKIT
Il en reste null

Comprend

Kit include steel drawer, built-in power frame, power strip, bottom & top Shelve, LED lights

The pre-configured components of Benchpro Bench Complete Kit make lab setup simple.

The pre-configured components of Bench Complete Kit make lab setup simple. FisherBrand Complete Kit include steel drawer, built-in power frame, power strip, bottom and top Shelve, LED lights. White Steel. 25 year unconditional manufacturer warranty

  • Pre-configured for simple setup
  • Steel drawer
  • Built-in power frame
  • Power strip
  • Bottom and top shelves
  • LED lights
  • White steel
  • 25 year unconditional warranty

Order Info

Kit does NOT include the workbench/table

Warranty

25-year manufacturer warranty

Spécifications

Product Type Bench Component Kit
Width (English) 48 in.
Width (Metric) 122 cm
Includes Steel drawer 10.1 + 15.2 + 30.4cm (4 + 6 + 12 in.), 4 outlets frame power, 8 outlets power strip, bottom Shelf, (1) ESD top shelf, (1) ESD top shelf with built-in LED light, 91.4cm (36 in.) powered upright, overhead LED light fixture