Learn More
BRANDTECH™ BRAND™ 48-Well Non-Skirted Microplates, BIO-CERT™ PCR QUALITY
Optimized for reliable and precise results in a wide range of PCR applications.
$426.72 - $475.09
Specifications
Barcode No Barcode
Binding Property Untreated
Certifications/Compliance RNase, DNase, DNA-free
Format Standard
Array 6 x 8
View More Specs
Products 2
Catalog Number Mfr. No. Color Price Quantity  
Catalog Number Mfr. No. Color Price Quantity  
13-878-204
View Documents
BRANDTECH™ BRAND™
781416
White
Pack of 20 Each for $475.09
Only null left
13-882-320
View Documents
BRANDTECH™ BRAND™
481415
Clear
Pack of 20 Each for $426.72
Only null left
Description
Description
BRAND™ PCR products deliver excellent, reproducible results for a wide range of PCR and qPCR applications. Manufactured from high-quality virgin resins, these products are free from lubricants, chemical additives, DNA, DNase, RNase, and other PCR inhibitors. Key features include:
  • Exceptional Quality and Purity
    • BIO-CERT™ PCR QUALITY ensures plates are free of human DNA, RNase, DNase, pyrogens, and PCR inhibitors
    • Free of contaminants that could interfere with reagents and samples
    • Ultra-smooth vessel interiors minimize enzyme nucleic acid binding
  • Precision Thermal Performance
    • Thin, uniform walls ensure consistent, precise thermal transfer for reliable results
    • Designed for rapid and accurate heat transfer, reducing cycle times
  • Wide Compatibility
    • Compact design is compatible with most standard thermal cyclers, ensuring convenient handling
    • White PCR plates, enhanced with titanium dioxide (TiO2), optimize fluorescence signal reflection and are recommended for qPCR applications
  • User-Friendly Features
    • Blue alphanumeric coding for easy sample identification
    • Well rims designed to prevent cross-contamination and ensure reliable sealing with tailored mats
Specifications
Specifications
No Barcode
RNase, DNase, DNA-free
6 x 8
Single-use
Non-skirted
0.2 mL
Non-sterile
Conical Bottom (V-well)
20/Pk.
Untreated
Standard
Autoclavable
Thermal Cyclers
Polypropylene
48
Non-treated
Yes
Yes