missing translation for 'onlineSavingsMsg'
Learn More

BRANDTECH™ BRAND™ 48-Well Non-Skirted Microplates, BIO-CERT™ PCR QUALITY

Catalog No. 13878204
Change view
Click to view available options
Color:
Clear
White
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
13878204 White
13882320 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13878204 Supplier BRANDTECH™ BRAND™ Supplier No. 781416
Only null left

Optimized for reliable and precise results in a wide range of PCR applications.

BRAND™ PCR products deliver excellent, reproducible results for a wide range of PCR and qPCR applications. Manufactured from high-quality virgin resins, these products are free from lubricants, chemical additives, DNA, DNase, RNase, and other PCR inhibitors. Key features include:

  • Exceptional Quality and Purity
    • BIO-CERT™ PCR QUALITY ensures plates are free of human DNA, RNase, DNase, pyrogens, and PCR inhibitors
    • Free of contaminants that could interfere with reagents and samples
    • Ultra-smooth vessel interiors minimize enzyme nucleic acid binding
  • Precision Thermal Performance
    • Thin, uniform walls ensure consistent, precise thermal transfer for reliable results
    • Designed for rapid and accurate heat transfer, reducing cycle times
  • Wide Compatibility
    • Compact design is compatible with most standard thermal cyclers, ensuring convenient handling
    • White PCR plates, enhanced with titanium dioxide (TiO2), optimize fluorescence signal reflection and are recommended for qPCR applications
  • User-Friendly Features
    • Blue alphanumeric coding for easy sample identification
    • Well rims designed to prevent cross-contamination and ensure reliable sealing with tailored mats

Specifications

Barcode No Barcode
Binding Property Untreated
Certifications/Compliance RNase, DNase, DNA-free
Format Standard
Array 6 x 8
Autoclavable Autoclavable
Color White
Disposable Single-use
Product Type qPCR 48-well plate
For Use With (Equipment) Thermal Cyclers
Skirt Style Non-skirted
Material Polypropylene
Volume (Metric) Well 0.2 mL
No. of Wells 48
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Conical Bottom (V-well)
Dnase Free Yes
For Use With (Application) RT-PCR, qPCR applications
No. per Pack 40
Quantity 20/Pk.
RNase Free Yes
Show More Show Less