missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ 40nm Colloidal Gold Nanoparticles 20 OD
Description
- Uniform Shape
- Low co-efficient of variation in terms of size
- High efficiency antibody binding for maximum sensitivity
- Available at high concentrations to simplify assay development
Specifications
Specifications
| Target | 40nm Colloidal Gold Nanoparticles 20 OD |
| Product Type | Antibody Labeling Kit |
| Content And Storage | Store at 4°C. Do not freeze. |
| Product Line | Novus |
| Quantity | 100 mL |