missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek 405 TS Touch Microplate Washer, IVD
GREENER_CHOICE

Catalog No. BT405TSRSI
Change view
Click to view available options
Model:
405TSHT
405TSHTV
405TSR
405TSRS
405TSRSQ
405TSRV
405TSRVS
405TSRVSQ
405TSU
405TSUS
405TSUSQ
405TSUV
405TSUVS
405TSUVSQ
Includes:
4-Buffer Switching, 96-well microplate washing
4-Buffer Switching, Ultrasonic Advantage, 96-/384-well microplate washing
4-Buffer Switching, Ultrasonic Advantage, 96-well microplate washing
4-Buffer Switching, Ultrasonic Advantage, 96-well microplate washing, Verify Technology
4-Buffer Switching, Ultrasonic Advantage, Faster 384-well microplate washing
4-Buffer Switching, Ultrasonic Advantage, Optimized Cell Washing, 96-/384-well microplate washing
4-Buffer Switching, Ultrasonic Advantage, Optimized Cell Washing, 96-/384-well microplate washing, Verify Technology
96-well microplate washing
Optimized Cell Washing, 96-/384-well microplate washing
Optimized Cell Washing, Ultrasonic Advantage, 96-/384-well microplate washing
Optimized Cell Washing, Ultrasonic Advantage, 96-/384-well microplate washing, Verify Technology
Ultrasonic Advantage, 96-well microplate washing
Ultrasonic Advantage, 96-well microplate washing, Verify Technology
Ultrasonic Advantage, Faster 384-well microplate washing
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Includes
BT405TSRSI 405TSR 96-well microplate washing
BT405TSRSSI 405TSRS Ultrasonic Advantage, 96-well microplate washing
BT405TSUSI 405TSU Optimized Cell Washing, 96-/384-well microplate washing
BT405TSHTSI 405TSHT Ultrasonic Advantage, Faster 384-well microplate washing
BT405TSRVSI 405TSRV 4-Buffer Switching, 96-well microplate washing
BT405TRSQSI 405TSRSQ Ultrasonic Advantage, 96-well microplate washing, Verify Technology
BT405TRVSSI 405TSRVS 4-Buffer Switching, Ultrasonic Advantage, 96-well microplate washing
BT405TSUSSI 405TSUS Optimized Cell Washing, Ultrasonic Advantage, 96-/384-well microplate washing
BT405TSUVSI 405TSUV 4-Buffer Switching, Ultrasonic Advantage, 96-/384-well microplate washing
BT405TUSQSI 405TSUSQ Optimized Cell Washing, Ultrasonic Advantage, 96-/384-well microplate washing, Verify Technology
BT405THTVSI 405TSHTV 4-Buffer Switching, Ultrasonic Advantage, Faster 384-well microplate washing
BT405TRVQSI 405TSRVSQ 4-Buffer Switching, Ultrasonic Advantage, 96-well microplate washing, Verify Technology
BT405TUVQSI 405TSUVSQ 4-Buffer Switching, Ultrasonic Advantage, Optimized Cell Washing, 96-/384-well microplate washing, Verify Technology
BT405TUVSSI 405TSUVS 4-Buffer Switching, Ultrasonic Advantage, Optimized Cell Washing, 96-/384-well microplate washing
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Catalog No. BT405TSRSI Supplier Agilent BioTek Supplier No. 405TSRSI

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The BioTek 405 TS washer has an excellent reputation in life science research for outstanding results in many applications. For In Vitro Diagnostic Use.

The BioTek 405 TS washer has an excellent reputation in life science research for outstanding results in many applications. The 405 TS has a unique set of features designed to efficiently wash cell-based assays, microsphere-based assays, and ELISA applications. Programming and operation via the touchscreen interface is intuitive and simple.

The patented Verify technology automatically checks for manifold tube blockages before a wash cycle, and Ultrasonic Advantage automatically cleans any blocked manifold tubes. These functions save time and eliminate frustration while ensuring successful results.

 

Must be purchased together with required dispense/waste system accessory for use. Please contact your Fisher Sales Representative for more information.

Specifications

Assays ELISA, Cell-based assays (model dependent), Magnetic and polystyrene bead, Multiplex assays, Bead-based ELISAs, Filtration-to-waste processes
Certifications/Compliance CE and TUV marked. RoHS compliant.
Connections USB (2)
Bottles Optional (4 or 10 L)
Precision <3% CV: 300 μL/well (96-well washing); <4% CV: 80 μL/well (384-well washing)
Dispense Volume 25 to 3000 μL/well (192-pin manifold); 50 to 3000 μL/well (96-pin manifold)
Plate Type 96- and 384-well, Low Profile and Standard, Solid and Filter Bottom (Option) Plates
Residual Volume ≤2 μL/well
Washing Speed 96-wells: 300 μL/well, 3 cycles: <30 s; 384-wells: 100 μL/well, 3 cycles: <80 s; 384-wells: 400 μL/well, 1 cycle: <20 s
Soak Time Programmable in minutes and seconds, up to 60 min.
Type Microplate Washer
Weight (Metric) With internal buffer switching - 16.5 kg; Without - 13.5 kg
Wash Cycles 1 to 250
Wash Volume 25 to 3000 μL/well
Display LED
Flow Rate High to Low Flow; Optimized rates for cell assays
Fluid Delivery Type Internal positive displacement pump
For Use With (Application) ELISA, MSD assays, HCS immune cytochemistry, FLIPR Ca2+ flux, Cell-based assays, Magnetic and polystyrene bead processing (gene expression assays, cytokine assays), ELISPOT assays
Frequency 50/60 Hz
Model 405TSR
Includes 96-well microplate washing
Width (English) 14 in.
Width (Metric) 35.6 cm
Height (English) 10 in.
Height (Metric) 25.4 cm
Depth (English) 17 in.
Depth (Metric) 43.2 cm
Automation Interface High Resolution LED Backlit Touch Screen
Weight (English) With internal buffer switching - 36 lb.; Without - 30 lb.
Voltage 100/240 V
Show More Show Less