Learn More
Agilent BioTek 405 TS Microplate Washer

The BioTek 405 TS washer has an excellent reputation in life science research for outstanding results in many applications.
Supplier: Agilent BioTek 405TSUVSN
Description
The BioTek 405 TS washer has an excellent reputation in life science research for outstanding results in many applications. The 405 TS has a unique set of features designed to efficiently wash cell-based assays, microsphere-based assays, and ELISA applications. Programming and operation via the touchscreen interface is intuitive and simple.
Must be purchased together with required dispense/waste system accessory for use. Please contact your Fisher Sales Representative for more information.
Chemical Identifiers
| CE and TUV marked. RoHS compliant. | |
| Optional (4 or 10 L) | |
| 96- and 384-well, Low Profile and Standard, Solid and Filter Bottom (Option) Plates | |
| Programmable in minutes and seconds, up to 60 min. | |
| With internal buffer switching - 16.5 kg; Without - 13.5 kg | |
| 25 to 3000 μL/well | |
| Internal positive displacement pump | |
| 14 in. | |
| 10 in. | |
| 17 in. | |
| High Resolution LED Backlit Touch Screen | |
| 100/240 V |
| USB (2) | |
| 25 to 3000 μL/well (192-pin manifold); 50 to 3000 μL/well (96-pin manifold) | |
| ≤2 μL/well | |
| Microplate Washer | |
| 1 to 250 | |
| LED | |
| 50/60 Hz | |
| 35.6 cm | |
| 25.4 cm | |
| 43.2 cm | |
| With internal buffer switching - 36 lb.; Without - 30 lb. |
Specifications
| ELISA, Cell-based assays (model dependent), Magnetic and polystyrene bead, Multiplex assays, Bead-based ELISAs, Filtration-to-waste processes | |
| USB (2) | |
| <3% CV: 300 μL/well (96-well washing); <4% CV: 80 μL/well (384-well washing) | |
| 96- and 384-well, Low Profile and Standard, Solid and Filter Bottom (Option) Plates | |
| 96-wells: 300 μL/well, 3 cycles: <30 s; 384-wells: 100 μL/well, 3 cycles: <80 s; 384-wells: 400 μL/well, 1 cycle: <20 s | |
| Microplate Washer | |
| 1 to 250 | |
| LED | |
| Internal positive displacement pump | |
| 50/60 Hz | |
| Automated Internal 4-Buffer Switching; 96-/384-well microplate washing | |
| 35.6 cm | |
| 25.4 cm | |
| 43.2 cm | |
| With internal buffer switching - 36 lb.; Without - 30 lb. |
| CE and TUV marked. RoHS compliant. | |
| Optional (4 or 10 L) | |
| 25 to 3000 μL/well (192-pin manifold); 50 to 3000 μL/well (96-pin manifold) | |
| ≤2 μL/well | |
| Programmable in minutes and seconds, up to 60 min. | |
| With internal buffer switching - 16.5 kg; Without - 13.5 kg | |
| 25 to 3000 μL/well | |
| High to Low Flow; Optimized rates for cell assays | |
| ELISA, MSD assays, HCS immune cytochemistry, FLIPR Ca2+ flux, Cell-based assays, Magnetic and polystyrene bead processing (gene expression assays, cytokine assays), ELISPOT assays | |
| 405TSUV | |
| 14 in. | |
| 10 in. | |
| 17 in. | |
| High Resolution LED Backlit Touch Screen | |
| 100/240 V |