missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek 405 TS Microplate Washer
GREENER_CHOICE

The BioTek 405 TS washer has an excellent reputation in life science research for outstanding results in many applications.

Supplier:  Agilent BioTek 405TSRSQSN

 View more versions of this product

Catalog No. BT405TSRSQ

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

The BioTek 405 TS washer has an excellent reputation in life science research for outstanding results in many applications. The 405 TS has a unique set of features designed to efficiently wash cell-based assays, microsphere-based assays, and ELISA applications. Programming and operation via the touchscreen interface is intuitive and simple.

The patented Verify technology automatically checks for manifold tube blockages before a wash cycle, and Ultrasonic Advantage automatically cleans any blocked manifold tubes. These functions save time and eliminate frustration while ensuring successful results.

 

Must be purchased together with required dispense/waste system accessory for use. Please contact your Fisher Sales Representative for more information.

Specifications

Chemical Identifiers

CE and TUV marked. RoHS compliant.
Optional (4 or 10 L)
96- and 384-well, Low Profile and Standard, Solid and Filter Bottom (Option) Plates
Programmable in minutes and seconds, up to 60 min.
With internal buffer switching - 16.5 kg; Without - 13.5 kg
25 to 3000 μL/well
Internal positive displacement pump
14 in.
10 in.
17 in.
High Resolution LED Backlit Touch Screen
100/240 V
USB (2)
25 to 3000 μL/well (192-pin manifold); 50 to 3000 μL/well (96-pin manifold)
≤2 μL/well
Microplate Washer
1 to 250
LED
50/60 Hz
35.6 cm
25.4 cm
43.2 cm
With internal buffer switching - 36 lb.; Without - 30 lb.

Specifications

ELISA, Cell-based assays (model dependent), Magnetic and polystyrene bead, Multiplex assays, Bead-based ELISAs, Filtration-to-waste processes
USB (2)
<3% CV: 300 μL/well (96-well washing); <4% CV: 80 μL/well (384-well washing)
96- and 384-well, Low Profile and Standard, Solid and Filter Bottom (Option) Plates
96-wells: 300 μL/well, 3 cycles: <30 s; 384-wells: 100 μL/well, 3 cycles: <80 s; 384-wells: 400 μL/well, 1 cycle: <20 s
Microplate Washer
1 to 250
LED
Internal positive displacement pump
50/60 Hz
Ultrasonic Advantage; 96-well microplate washing; Verify Technology
35.6 cm
25.4 cm
43.2 cm
With internal buffer switching - 36 lb.; Without - 30 lb.
CE and TUV marked. RoHS compliant.
Optional (4 or 10 L)
25 to 3000 μL/well (192-pin manifold); 50 to 3000 μL/well (96-pin manifold)
≤2 μL/well
Programmable in minutes and seconds, up to 60 min.
With internal buffer switching - 16.5 kg; Without - 13.5 kg
25 to 3000 μL/well
High to Low Flow; Optimized rates for cell assays
ELISA, MSD assays, HCS immune cytochemistry, FLIPR Ca2+ flux, Cell-based assays, Magnetic and polystyrene bead processing (gene expression assays, cytokine assays), ELISPOT assays
405TSRSQ
14 in.
10 in.
17 in.
High Resolution LED Backlit Touch Screen
100/240 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions