Learn More
Agilent BioTek 405LS Microplate Washer, IVD

The BioTek 405 LS washer has a well earned reputation as an economical plate washer that offers excellent performance and reliability. For In Vitro Diagnostic Use.
$17,530.60 - $45,477.91
Specifications
| Certifications/Compliance | CE and TUV marked. RoHS compliant. |
|---|---|
| Bottles | Optional (4 or 10 L) |
| Precision | <3% CV: 300 μL/well (96-well washing); <4% CV: 80 μL/well (384-well washing) |
| Dispense Volume | 25 to 3000 μL/well |
| Plate Type | 96- and 384-well, Low Profile and Standard, Solid and Filter Bottom (Pore Sizes 0.45 to 1.2 μm) Plates |
| Catalog Number | Mfr. No. | Model | Includes | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Model | Includes | Price | Quantity & Availability | ||||
BT405LSRSI
![]() |
Agilent BioTek
405LSRSI |
405LSR | 96-well microplate washing |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BT405LSUSI
![]() |
Agilent BioTek
405LSUSI |
405LSU | 96-/384-well microplate washing |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BT405LSHTSI
![]() |
Agilent BioTek
405LSHTSI |
405LSHT | Ultrasonic Advantage; Faster 384-well microplate washing |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BT405LSRSSI
![]() |
Agilent BioTek
405LSRSSI |
405LSRS | Ultrasonic Advantage; 96-well microplate washing |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BT405LSUSSI
![]() |
Agilent BioTek
405LSUSSI |
405LSUS | Ultrasonic Advantage; 96-/384-well microplate washing |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BT405LSRVSI
![]() |
Agilent BioTek
405LSRVSI |
405LSRV | Automated Internal 4-Buffer Switching; 96-well microplate washing |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BT405LSUVSI
![]() |
Agilent BioTek
405LSUVSI |
405LSUV | Automated Internal 4-Buffer Switching; 96-/384-well microplate washing |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BT405LUSQSI
![]() |
Agilent BioTek
405LSUSQSI |
405LSUSQ | Ultrasonic Advantage; 96-/384-well microplate washing; Verify Technology |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BT405LRSQSI
![]() |
Agilent BioTek
405LSRSQSI |
405LSRSQ | Ultrasonic Advantage; 96-well microplate washing; Verify Technology |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BT405LHTVSI
![]() |
Agilent BioTek
405LSHTVSI |
405LSHTV | Automated Internal 4-Buffer Switching; Faster 384-well microplate washing |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BT405LRVSSI
![]() |
Agilent BioTek
405LSRVSSI |
405LSRVS | Automated Internal 4-Buffer Switching; Ultrasonic Advantage; 96-well microplate washing |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BT405LUVSSI
![]() |
Agilent BioTek
405LSUVSSI |
405LSUVS | Automated Internal 4-Buffer Switching; Ultrasonic Advantage; 96-/384-well microplate washing |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BT405LRVQSI
![]() |
Agilent BioTek
405LSRVSQSI |
405LSRVSQ | Automated Internal 4-Buffer Switching; Ultrasonic Advantage; 96-well microplate washing; Verify Technology |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BT405LUVQSI
![]() |
Agilent BioTek
405LSUVSQSI |
405LSUVSQ | Automated Internal 4-Buffer Switching; Ultrasonic Advantage; 96-/384-well microplate washing; Verify Technology |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
The BioTek 405 LS washer has a well earned reputation as an economical plate washer that offers excellent performance and reliability. Washing protocols range from gentle cell washing to vigorous ELISA workflows. The 405 LS has a simple user interface for creation and execution of wash, dispense, aspirate, and maintenance protocols within the extensive onboard software
For use under computer control, Agilent BioTek LHC software used with the 405 LS is available in standard and 21 CFR Part 11 compliant versions. The optional Agilent BioTek BioStack is available for walk-away automation of up to 50 microplates.
Must be purchased together with required dispense/waste system accessory for use. Please contact your Fisher Sales Representative for more information.
Specifications
| CE and TUV marked. RoHS compliant. | |
| <3% CV: 300 μL/well (96-well washing); <4% CV: 80 μL/well (384-well washing) | |
| 96- and 384-well, Low Profile and Standard, Solid and Filter Bottom (Pore Sizes 0.45 to 1.2 μm) Plates | |
| Programmable in minutes and seconds, up to 60 min. | |
| With internal buffer switching - 16.5 kg; Without - 13.5 kg | |
| High to Low Flow; Optimized rates for cell assays | |
| 50/60 Hz | |
| 14 in. | |
| 10 in. | |
| 17 in. | |
| With internal buffer switching - 36 lb.; Without - 30 lb. |
| Optional (4 or 10 L) | |
| 25 to 3000 μL/well | |
| 96-wells: 300 μL/well, 3 cycles: <30 s; 384-wells: 100 μL/well, 3 cycles: <80 s; 384-wells: 400 μL/well, 1 cycle: <20 s | |
| Microplate Washer | |
| 1 to 250 | |
| 10 to 85%, noncondensing | |
| 1 μL | |
| 35.6 cm | |
| 25.4 cm | |
| 43.2 cm | |
| 100/240 V |