missing translation for 'onlineSavingsMsg'
Learn More

4-Vinylpyridine (stabilized with HQ) 95.0+%, TCI America™

Catalog No. V015025ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V015025ML 25 mL
V0150500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. V015025ML Supplier TCI America Supplier No. V015025ML

Chemical Identifiers

CAS 100-43-6
Molecular Formula C7H7N
Molecular Weight (g/mol) 105.14
MDL Number MFCD00006447
InChI Key KFDVPJUYSDEJTH-UHFFFAOYSA-N
Synonym 4-vinylpyridine, pyridine, 4-ethenyl, pyridine, 4-vinyl, gamma-vinylpyridine, unii-i56g67xm8d, ccris 5240, 4vp, 4-pyridylethylene, 4-vinyl-pyridine, 4-vinyl pyridine
PubChem CID 7502
IUPAC Name 4-ethenylpyridine
SMILES C=CC1=CC=NC=C1

Specifications

Color Yellow
Boiling Point 59°C
Quantity 25 mL
UN Number 3073
Formula Weight 105.14
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Vinylpyridine (stabilized with HQ)
EINECSNumber (5)-0717
RTECSNumber UU1045000
TSCA Yes
Recommended Storage Refrigerator