missing translation for 'onlineSavingsMsg'
Learn More

4-Vinylphenyl Acetate (stabilized with TBC) 98.0+%, TCI America™

Catalog No. p-7117508
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15515G 5 g
A155125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A15515G Supplier TCI America Supplier No. A15515G
Only null left

Chemical Identifiers

CAS 2628-16-2
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.188
MDL Number MFCD00075734
InChI Key JAMNSIXSLVPNLC-UHFFFAOYSA-N
Synonym 4-acetoxystyrene, 4-vinylphenyl acetate, 4-ethenylphenol acetate, p-acetoxystyrene, phenol, 4-ethenyl-, acetate, p-vinylphenol acetate, unii-7s21904ayn, phenol, 4-ethenyl-, 1-acetate, 4-ethenylphenyl acetate, acetic acid 4-vinylphenyl ester
PubChem CID 75821
IUPAC Name (4-ethenylphenyl) acetate
SMILES CC(=O)OC1=CC=C(C=C1)C=C

Specifications

Color Yellow
Quantity 5 g
Formula Weight 162.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Vinylphenyl Acetate (stabilized with TBC)
EINECSNumber (3)-2784
RTECSNumber SL3784000
TSCA Yes