missing translation for 'onlineSavingsMsg'
Learn More

4-Vinylphenol, min 10% soln. in propylene glycol

Catalog No. AAL1090203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1090203 1 g
AAL1090206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1090203 Supplier Thermo Scientific Chemicals Supplier No. L1090203

CAS: 2628-17-3 | C8H8O | 120.151 g/mol

4-Vinylphenol acts as a monomer and used in the preparation of poly-4-vinylphenol polymer, which is used in electronics as a dielectric layer in organic transistors in organic thin-film-transistor liquid-crystal displays (TFT LCD). It is a phenolic compound and presents in wine and beer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Vinylphenol acts as a monomer and used in the preparation of poly-4-vinylphenol polymer, which is used in electronics as a dielectric layer in organic transistors in organic thin-film-transistor liquid-crystal displays (TFT LCD). It is a phenolic compound and presents in wine and beer.

Solubility
Slightly miscible with water.

Notes
Hygroscopic. Moisture and heat sensitive. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 2628-17-3
Molecular Formula C8H8O
Molecular Weight (g/mol) 120.151
MDL Number MFCD00017593
InChI Key FUGYGGDSWSUORM-UHFFFAOYSA-N
Synonym 4-vinylphenol, 4-hydroxystyrene, p-vinylphenol, p-hydroxystyrene, phenol, 4-ethenyl, 4-vinylphenol, 10 wt.% in propylene glycol, phenol, p-vinyl, unii-oa7v1sm8yl, fema no. 3739, poly 4-hydroxystyrene
PubChem CID 62453
ChEBI CHEBI:1883
IUPAC Name 4-ethenylphenol
SMILES C=CC1=CC=C(C=C1)O

Specifications

Density 1.04
Flash Point >100°C (212°F)
Refractive Index 1.447
Quantity 1 g
Beilstein 506844
Sensitivity Hygroscopic
Solubility Information Slightly miscible with water.
Formula Weight 120.15
Concentration or Composition (by Analyte or Components) ≥10% soln. in propylene glycol
Chemical Name or Material 4-Vinylphenol
Hazard Category H341-H361-H372
Hazard Statement GHS H Statement
H318
Causes serious eye damage.
Precautionary Statement P201-P202-P260-P264b-P270-P281-P308+P313-P501c
EINECSNumber 220-103-6
RTECSNumber SN3800000
TSCA No
Recommended Storage Store at -20°C; Store under Nitrogen

RUO – Research Use Only