missing translation for 'onlineSavingsMsg'
Learn More

4-Vinylbiphenyl 98.0+%, TCI America™

Catalog No. p-7137904
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V00541G 1 g
V00545G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. V00541G Supplier TCI America Supplier No. V00541G

Chemical Identifiers

CAS 2350-89-2
Molecular Formula C14H12
Molecular Weight (g/mol) 180.25
MDL Number MFCD00008620
InChI Key HDBWAWNLGGMZRQ-UHFFFAOYSA-N
Synonym 4-vinylbiphenyl, p-vinylbiphenyl, 4-phenylstyrene, biphenyl, 4-vinyl, 1,1'-biphenyl, 4-ethenyl, p-phenylstyrene, 4-ethenyl-1,1'-biphenyl, 4-vinyldiphenyl, 4-vinyl biphenyl, 4-vinyl-biphenyl
PubChem CID 16883
IUPAC Name 4-ethenyl-1,1'-biphenyl
SMILES C=CC1=CC=C(C=C1)C1=CC=CC=C1

Specifications

Melting Point 121°C
Color White-Yellow
Quantity 1 g
Formula Weight 180.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Vinylbiphenyl
TSCA No
Recommended Storage Refrigerator