accessibility menu, dialog, popup

UserName

CAS RN 2350-89-2

IUPAC Name: 4-ethenyl-1,1'-biphenyl
Synonyms: 4-ethenyl-1,1'-biphenyl
Molecular Weight (g/mol): 180.25
Molecular Formula: C14H12
InChi Key: HDBWAWNLGGMZRQ-UHFFFAOYSA-N
SMILES: C=CC1=CC=C(C=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)