missing translation for 'onlineSavingsMsg'
Learn More

4-Vinylbenzylamine (stabilized with MEHQ) 92.0+%, TCI America™

Catalog No. V01011G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V01011G 1 g
V01015G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. V01011G Supplier TCI America Supplier No. V01011G
Only null left

Chemical Identifiers

CAS 50325-49-0
Molecular Formula C9H11N
Molecular Weight (g/mol) 133.194
MDL Number MFCD08704374
InChI Key WAVDSLLYAQBITE-UHFFFAOYSA-N
Synonym 4-(Aminomethyl)styrene
PubChem CID 11073466
IUPAC Name (4-ethenylphenyl)methanamine
SMILES C=CC1=CC=C(C=C1)CN

Specifications

Color Yellow
Boiling Point 60°C
Quantity 1 g
Formula Weight 133.19
Percent Purity ≥92.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Vinylbenzylamine (stabilized with MEHQ)
TSCA No
Recommended Storage Freezer