Learn More
4-Vinylbenzyl chloride, 90%, technical
CAS: 1592-20-7 | C9H9Cl | 152.62 g/mol
$319.37 - $319.37
Chemical Identifiers
| CAS | 1592-20-7 |
|---|---|
| Molecular Formula | C9H9Cl |
| Molecular Weight (g/mol) | 152.62 |
| MDL Number | MFCD00051362 |
| InChI Key | ZRZHXNCATOYMJH-UHFFFAOYSA-N |
| Synonym | 4-vinylbenzyl chloride, 1-chloromethyl-4-vinylbenzene, 4-chloromethylstyrene, 4-vinylbenzylchloride, benzene, 1-chloromethyl-4-ethenyl, 1-chloromethyl-4-ethenylbenzene, 4-chloromethyl styrene, p-vinylbenzyl chloride, unii-cjl4oj6bs0, cjl4oj6bs0 |
| PubChem CID | 74126 |
| IUPAC Name | 1-(chloromethyl)-4-ethenylbenzene |
| SMILES | ClCC1=CC=C(C=C)C=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Qty | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Qty | ||||
|
AC422521000
|
Thermo Scientific Chemicals
422521000 |
100 g | Glass bottle |
Each for $319.37
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1592-20-7 | |
| 152.62 | |
| ZRZHXNCATOYMJH-UHFFFAOYSA-N | |
| 74126 | |
| ClCC1=CC=C(C=C)C=C1 |
| C9H9Cl | |
| MFCD00051362 | |
| 4-vinylbenzyl chloride, 1-chloromethyl-4-vinylbenzene, 4-chloromethylstyrene, 4-vinylbenzylchloride, benzene, 1-chloromethyl-4-ethenyl, 1-chloromethyl-4-ethenylbenzene, 4-chloromethyl styrene, p-vinylbenzyl chloride, unii-cjl4oj6bs0, cjl4oj6bs0 | |
| 1-(chloromethyl)-4-ethenylbenzene |
Specifications
| 1592-20-7 | |
| 100.0 | |
| 1.0800g/mL | |
| Authentic | |
| Glass bottle | |
| 1.5731 to 1.5751 | |
| MFCD00051362 | |
| 1.08 | |
| ZRZHXNCATOYMJH-UHFFFAOYSA-N | |
| 1-(chloromethyl)-4-ethenylbenzene | |
| about 500ppm 4-tert-Butylcatechol, about 500ppm Nitrocresol + Nitrophenol | |
| 152.62 | |
| Technical | |
| 4-Vinylbenzyl chloride |
| 88.0 | |
| Yellow | |
| 105°C | |
| 88% min. (GC) | |
| C9H9Cl | |
| CH2CHC6H4CH2Cl | |
| 100 g | |
| 4-vinylbenzyl chloride, 1-chloromethyl-4-vinylbenzene, 4-chloromethylstyrene, 4-vinylbenzylchloride, benzene, 1-chloromethyl-4-ethenyl, 1-chloromethyl-4-ethenylbenzene, 4-chloromethyl styrene, p-vinylbenzyl chloride, unii-cjl4oj6bs0, cjl4oj6bs0 | |
| ClCC1=CC=C(C=C)C=C1 | |
| 152.62 | |
| 74126 | |
| 90% | |
| Liquid |
Safety and Handling
GHS H Statement
Harmful in contact with skin.
May cause an allergic skin reaction.
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 216-471-2
RUO – Research Use Only