missing translation for 'onlineSavingsMsg'
Learn More

4-Vinylbenzeneboronic acid, 97%

Catalog No. p-7027165
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2370903 1 g
AAB2370906 5 g
AAB2370914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2370903 Supplier Thermo Scientific Chemicals Supplier No. B2370903
Only null left

CAS: 2156-04-9 | C8H9BO2 | 147.97 g/mol

4-Vinylbenzeneboronic acid is widely used in Suzuki reaction. It is also used in the preparation of boronic acid containing polymers, which is used in potential biomedical applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Vinylbenzeneboronic acid is widely used in Suzuki reaction. It is also used in the preparation of boronic acid containing polymers, which is used in potential biomedical applications.

Solubility
Soluble in ethanol, dimethylsulfoxide, tetrahydrofuran and diethyl ether. Slightly soluble in water.

Notes
Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 2156-04-9
Molecular Formula C8H9BO2
Molecular Weight (g/mol) 147.97
MDL Number MFCD00239441
InChI Key QWMJEUJXWVZSAG-UHFFFAOYSA-N
Synonym 4-vinylphenylboronic acid, 4-vinylbenzeneboronic acid, 4-vinylphenyl boronic acid, 4-ethenylphenyl boronic acid, 4-vinylboronic acid, p-styrylboronic acid, p-vinylphenylboronic acid, styrene-4-boronic acid, boronic acid, 4-ethenylphenyl, 4-styreneboronic acid
PubChem CID 2734393
IUPAC Name (4-ethenylphenyl)boronic acid
SMILES OB(O)C1=CC=C(C=C)C=C1

Specifications

Melting Point ∼190°C
Quantity 1 g
Beilstein 2829387
Solubility Information Soluble in ethanol,dimethylsulfoxide,tetrahydrofuran and diethyl ether. Slightly soluble in water.
Formula Weight 147.97
Percent Purity 97%
Chemical Name or Material 4-Vinylbenzeneboronic acid
Hazard Category H302-H312-H315-H319-H332-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only