missing translation for 'onlineSavingsMsg'
Learn More

4-Vinylaniline, 90%, technical, stabilized

Catalog No. AC308480050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC308480050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC308480050 Supplier Thermo Scientific Chemicals Supplier No. 308480050
Only null left

CAS: 1520-21-4 | C8H9N | 119.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1520-21-4
Molecular Formula C8H9N
Molecular Weight (g/mol) 119.17
MDL Number MFCD00015329
InChI Key LBSXSAXOLABXMF-UHFFFAOYSA-N
Synonym 4-vinylaniline, 4-aminostyrene, p-aminostyrene, benzenamine, 4-ethenyl, 4-aminostryene, ccris 4326, benzenamine,4-ethenyl, p-vinylaniline, p-amino styrene, para-aminostyrene
PubChem CID 73700
IUPAC Name 4-ethenylaniline
SMILES C=CC1=CC=C(C=C1)N

Specifications

CAS Min % 88.0
CAS Max % 100.0
Melting Point 23°C to 24°C
Color Red to Yellow
Density 1.0100g/mL
Boiling Point 249°C
Flash Point 99°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Refractive Index 1.6250 to 1.629
Linear Formula H2C=CHC6H4NH2
Quantity 5 g
Specific Gravity 1.01
Formula Weight 119.17
Percent Purity 90%
Grade Technical
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 4-Vinylaniline
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
May cause damage to organs through prolonged or repeated exposure.
May cause respiratory irritation.
Suspected of causing cancer
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unw
EINECSNumber 216-185-8

RUO – Research Use Only