missing translation for 'onlineSavingsMsg'
Learn More

4-Vinyl-1-cyclohexene, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. p-7187535
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019952 5 mL
111018409 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111019952 Supplier Merck Emd Millipore Supplier No. 949505ML
Only null left

4-Vinyl-1-cyclohexene can be synthesized via cyclo-dimerization of butadiene in the presence of nitrosyl iron catalysts like [Fe(NO)2(CO)2] or [{FeCl(NO)2}2]/reductant.

Featured Industries: Environmental and Petroleum

Specifications

CAS 100-40-3
Ignition Point 517°F
Vapor Pressure 10.2 mmHg (25°C)
Vapor Density 3.76 (vs. air)
Density 0.831 g/mL (at 20°C)
Flash Point 16°C
Physical Form Neat
Linear Formula C6H9 CH=CH2
MDL Number MFCD00001576
Melting Point -101°C
Boiling Point 126°C to 127°C
Synonym 4-Ethenyl-1-cyclohexene; NSC 15760
Type Analytical Standard
Quantity 5 mL
Formula Weight 108.18
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less