missing translation for 'onlineSavingsMsg'
Learn More

4-Vinyl-1,3-dioxolan-2-one 98.0+%, TCI America™

Catalog No. V011425G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V011425G 25 g
V0114100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. V011425G Supplier TCI America Supplier No. V011425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4427-96-7
Molecular Formula C5H6O3
Molecular Weight (g/mol) 114.1
MDL Number MFCD00143315
InChI Key BJWMSGRKJIOCNR-UHFFFAOYSA-N
Synonym 4-vinyl-1,3-dioxolan-2-one, vinylethylene carbonate, 4-vinyl-1,3-dioxolan-2-one, stabilized with hydroquinone, 1,3-dioxolan-2-one, 4-ethenyl, acmc-1ajgp, vec;, ksc383e5n, 2-oxo-4-vinyl-1,3-dioxolane, 4-vinyl-1,3 dioxolan-2-one, 4-vinyl-1,3-dioxaolane-2-one
PubChem CID 2735038
IUPAC Name 4-ethenyl-1,3-dioxolan-2-one
SMILES C=CC1COC(=O)O1

Specifications

Color Colorless
Boiling Point 122°C
Quantity 25 g
UN Number 2810
Formula Weight 114.10
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Vinyl-1,3-dioxolan-2-one
EINECSNumber (5)-6745
TSCA Yes
Recommended Storage Refrigerator