accessibility menu, dialog, popup

UserName

CH2(S)OOO

CAS RN 4427-96-7

IUPAC Name: (4S)-4-ethenyl-1,3-dioxolan-2-one
Synonyms: (4S)-4-ethenyl-1,3-dioxolan-2-one
Molecular Weight (g/mol): 114.1
Molecular Formula: C5H6O3
InChi Key: BJWMSGRKJIOCNR-BYPYZUCNSA-N
SMILES: C=C[C@H]1COC(=O)O1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)