missing translation for 'onlineSavingsMsg'
Learn More

4-Vinyl-1,3-dioxolan-2-one, 99%

Catalog No. p-7040918
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6082218 50 g
AAH6082230 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6082218 Supplier Thermo Scientific Chemicals Supplier No. H6082218
Only null left

CAS: 4427-96-7 | C5H6O3 | 114.1 g/mol

It is used to synthesize functional polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used to synthesize functional polymers.

Solubility
Insoluble in water.

Notes
Store at 2-8°C. Incompatible with oxidizing agents and heat.

Chemical Identifiers

CAS 4427-96-7
Molecular Formula C5H6O3
Molecular Weight (g/mol) 114.1
MDL Number MFCD00143315
InChI Key BJWMSGRKJIOCNR-UHFFFAOYSA-N
Synonym 4-vinyl-1,3-dioxolan-2-one, vinylethylene carbonate, 4-vinyl-1,3-dioxolan-2-one, stabilized with hydroquinone, 1,3-dioxolan-2-one, 4-ethenyl, acmc-1ajgp, vec;, ksc383e5n, 2-oxo-4-vinyl-1,3-dioxolane, 4-vinyl-1,3 dioxolan-2-one, 4-vinyl-1,3-dioxaolane-2-one
PubChem CID 2735038
IUPAC Name 4-ethenyl-1,3-dioxolan-2-one
SMILES C=CC1COC(=O)O1

Specifications

Density 1.188
Boiling Point 237°C
Flash Point 96°C (205°F)
Refractive Index 1.45
Quantity 50 g
UN Number UN2810
Solubility Information Insoluble in water.
Formula Weight 114.1
Percent Purity 99%
Chemical Name or Material 4-Vinyl-1,3-dioxolan-2-one
Hazard Category H301
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only