missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethylthio)benzylamine 97.0+%, TCI America™

Supplier:  TCI America T27635G

 View more versions of this product

Catalog No. T27635G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

4-(Trifluoromethylthio)benzylamine
Colorless
C8H8F3NS
5 g
4-trifluoromethylthio benzylamine, 4-trifluoromethyl thio phenyl methanamine, 4-trifluoromethylsulfanyl phenyl methanamine, 4-trifluoromethyl sulfanyl phenyl methanamine, 1-4-trifluoromethyl thio phenyl methanamine, 1-4-trifluoromethyl sulfanyl phenyl methanamine, acmc-1chvc, rarechem al bw 0450, 4-trifluomethylthio benzylamine
C1=CC(=CC=C1CN)SC(F)(F)F
207.214
207.21
Liquid
128273-56-3
225°C
MFCD00190129
2735
LACURGWEZCFLBO-UHFFFAOYSA-N
[4-(trifluoromethylsulfanyl)phenyl]methanamine
2777875
≥97.0% (GC,T)

Chemical Identifiers

128273-56-3
207.214
LACURGWEZCFLBO-UHFFFAOYSA-N
2777875
C1=CC(=CC=C1CN)SC(F)(F)F
C8H8F3NS
MFCD00190129
4-trifluoromethylthio benzylamine, 4-trifluoromethyl thio phenyl methanamine, 4-trifluoromethylsulfanyl phenyl methanamine, 4-trifluoromethyl sulfanyl phenyl methanamine, 1-4-trifluoromethyl thio phenyl methanamine, 1-4-trifluoromethyl sulfanyl phenyl methanamine, acmc-1chvc, rarechem al bw 0450, 4-trifluomethylthio benzylamine
[4-(trifluoromethylsulfanyl)phenyl]methanamine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions