missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethylthio)benzoyl Chloride 98.0+%, TCI America™

Catalog No. p-7137389
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T27455G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T27455G Supplier TCI America Supplier No. T27455G
Only null left

Chemical Identifiers

CAS 330-14-3
Molecular Formula C8H4ClF3OS
Molecular Weight (g/mol) 240.624
MDL Number MFCD01631632
InChI Key BCMFTOCCLOAGBP-UHFFFAOYSA-N
Synonym 4-trifluoromethylthio benzoyl chloride, 4-trifluoromethyl thio benzoyl chloride, 4-trifluoromethylsulfanyl benzoyl chloride, 4-trifluoromethyl sulfanyl benzoyl chloride, 4-trifluomethylthio benzoyl chloride, benzoyl chloride, 4-trifluoromethyl thio, 4-trifluoromethyl sulphanyl benzoyl chloride, 4-trifluoromethylsulfanyl-benzoyl chloride, acmc-209hxt, 4-trifluoromethylthiobenzoylchloride
PubChem CID 2777865
IUPAC Name 4-(trifluoromethylsulfanyl)benzoyl chloride
SMILES C1=CC(=CC=C1C(=O)Cl)SC(F)(F)F

Specifications

Color Yellow
Boiling Point 230°C
Quantity 5 g
UN Number 2920
Formula Weight 240.62
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-(Trifluoromethylthio)benzoyl Chloride