missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Trifluoromethylthio)benzaldehyde 90.0+%, TCI America™
Supplier: TCI America T240525G
Specifications
| 4-(Trifluoromethylthio)benzaldehyde | |
| White-Yellow | |
| C8H5F3OS | |
| 25 g | |
| OPAXHVNZGUGCKW-UHFFFAOYSA-N | |
| 4-[(trifluoromethyl)sulfanyl]benzaldehyde | |
| 2777846 | |
| ≥90.0% (GC) |
| 4021-50-5 | |
| 85°C | |
| MFCD00129191 | |
| 4-trifluoromethylthio benzaldehyde, 4-trifluoromethyl thio benzaldehyde, 4-trifluoromethyl sulfanyl benzaldehyde, benzaldehyde, 4-trifluoromethyl thio, acmc-1agrc, ksc582m6p, 4-trifluomethylthio benzoldehyde, rarechem gt hw 0381 | |
| FC(F)(F)SC1=CC=C(C=O)C=C1 | |
| 206.18 | |
| 206.18 | |
| Crystalline Lumps |
Chemical Identifiers
| 4021-50-5 | |
| 206.18 | |
| OPAXHVNZGUGCKW-UHFFFAOYSA-N | |
| 2777846 | |
| FC(F)(F)SC1=CC=C(C=O)C=C1 |
| C8H5F3OS | |
| MFCD00129191 | |
| 4-trifluoromethylthio benzaldehyde, 4-trifluoromethyl thio benzaldehyde, 4-trifluoromethyl sulfanyl benzaldehyde, benzaldehyde, 4-trifluoromethyl thio, acmc-1agrc, ksc582m6p, 4-trifluomethylthio benzoldehyde, rarechem gt hw 0381 | |
| 4-[(trifluoromethyl)sulfanyl]benzaldehyde |
Safety and Handling
TSCA : No