missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethylthio)benzaldehyde 90.0+%, TCI America™

Catalog No. T24055G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T24055G 5 g
T240525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T24055G Supplier TCI America Supplier No. T24055G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4021-50-5
Molecular Formula C8H5F3OS
Molecular Weight (g/mol) 206.18
MDL Number MFCD00129191
InChI Key OPAXHVNZGUGCKW-UHFFFAOYSA-N
Synonym 4-trifluoromethylthio benzaldehyde, 4-trifluoromethyl thio benzaldehyde, 4-trifluoromethyl sulfanyl benzaldehyde, benzaldehyde, 4-trifluoromethyl thio, acmc-1agrc, ksc582m6p, 4-trifluomethylthio benzoldehyde, rarechem gt hw 0381
PubChem CID 2777846
IUPAC Name 4-[(trifluoromethyl)sulfanyl]benzaldehyde
SMILES FC(F)(F)SC1=CC=C(C=O)C=C1

Specifications

Color White-Yellow
Boiling Point 85°C
Quantity 5 g
Formula Weight 206.18
Percent Purity ≥90.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4-(Trifluoromethylthio)benzaldehyde