missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethylthio)benzaldehyde, 90+%

Catalog No. AAB2031303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2031303 1 g
AAB2031306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2031303 Supplier Thermo Scientific Chemicals Supplier No. B2031303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4021-50-5 | C8H5F3OS | 206.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4021-50-5
Molecular Formula C8H5F3OS
Molecular Weight (g/mol) 206.18
MDL Number MFCD00129191
InChI Key OPAXHVNZGUGCKW-UHFFFAOYSA-N
Synonym 4-trifluoromethylthio benzaldehyde, 4-trifluoromethyl thio benzaldehyde, 4-trifluoromethyl sulfanyl benzaldehyde, benzaldehyde, 4-trifluoromethyl thio, acmc-1agrc, ksc582m6p, 4-trifluomethylthio benzoldehyde, rarechem gt hw 0381
PubChem CID 2777846
IUPAC Name 4-(trifluoromethylsulfanyl)benzaldehyde
SMILES FC(F)(F)SC1=CC=C(C=O)C=C1

Specifications

Melting Point 29°C to 31°C
Density 1.31
Boiling Point 219°C
Flash Point 96°C (204°F)
Odor Stench
Refractive Index 1.511
Quantity 1 g
Beilstein 2328411
Sensitivity Air Sensitive
Formula Weight 206.19
Percent Purity ≥90%
Chemical Name or Material 4-(Trifluoromethylthio)benzaldehyde
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only