missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethylthio)aniline 98.0+%, TCI America™

Catalog No. p-7137409
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T27755G 5 g
T277525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T27755G Supplier TCI America Supplier No. T27755G

Chemical Identifiers

CAS 372-16-7
Molecular Formula C7H6F3NS
Molecular Weight (g/mol) 193.19
MDL Number MFCD00040926
InChI Key OHHHTUXVBNGOGI-UHFFFAOYSA-N
Synonym 4-trifluoromethylthio aniline, 4-trifluoromethyl thio aniline, 4-trifluoromethyl sulfanyl aniline, 4-aminophenyl trifluoromethyl sulfide, 4-aminophenyl trifluoromethyl sulphide, 4-trifluoromethylsulfanyl aniline, benzenamine, 4-trifluoromethyl thio, 4-trifluoromethylthio phenylamine
PubChem CID 123054
IUPAC Name 4-[(trifluoromethyl)sulfanyl]aniline
SMILES NC1=CC=C(SC(F)(F)F)C=C1

Specifications

Color Yellow
Boiling Point 103°C
Quantity 5 g
Formula Weight 193.19
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-(Trifluoromethylthio)aniline