missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethylsulfonyl)phenylacetic acid, 98%

Catalog No. AAH3229506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3229506 5 g
AAH3229503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3229506 Supplier Thermo Scientific Chemicals Supplier No. H3229506

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1099597-82-6 | C9H7F3O4S | 268.206 g/mol

4-(Trifluoromethylsulfonyl)phenylacetic acid, is used as a primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Trifluoromethylsulfonyl)phenylacetic acid, is used as a primary and secondary intermediate.

Solubility
Slightly soluble in water, soluble in organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 1099597-82-6
Molecular Formula C9H7F3O4S
Molecular Weight (g/mol) 268.206
MDL Number MFCD04973012
InChI Key FIEUCHICQSJBAU-UHFFFAOYSA-N
Synonym 4-trifluoromethylsulfony phenylacetic acid, 2-4-trifluoromethyl sulfonyl phenyl acetic acid, 4-trifluoromethanesulfonylphenyl acetic acid, 4-trifluoromethylsulfonyl phenylacetic acid, 4-trifluoromethanesulfonyl phenyl acetic acid
PubChem CID 40427164
IUPAC Name 2-[4-(trifluoromethylsulfonyl)phenyl]acetic acid
SMILES C1=CC(=CC=C1CC(=O)O)S(=O)(=O)C(F)(F)F

Specifications

Melting Point 125°C to 128°C
Quantity 5 g
Solubility Information Slightly soluble in water,soluble in organic solvents.
Formula Weight 268.21
Percent Purity 98%
Chemical Name or Material 4-(Trifluoromethylsulfonyl)phenylacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only