missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethylsulfonyl)benzonitrile, 97+%

Catalog No. p-7026728
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2290703 1 g
AAB2290706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2290703 Supplier Thermo Scientific Chemicals Supplier No. B2290703
Only null left

CAS: 312-21-0 | C8H4F3NO2S | 235.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 312-21-0
Molecular Formula C8H4F3NO2S
Molecular Weight (g/mol) 235.18
MDL Number MFCD01631631
InChI Key UHCGSMMNICMDIX-UHFFFAOYSA-N
Synonym 4-trifluoromethylsulfonyl benzonitrile, 4-trifluoromethyl sulfonyl benzonitrile, 4-trifluoromethanesulfonyl benzonitrile, 4-trifluoromethanesulfonylbenzonitrile, 4-trifluoromethyl sulfonyl benzenecarbonitrile, acmc-1clj3, 4-trifluoromethanesulphonylbenzonitrile, 4-trifluoromethylsulfonyl benzonitril
PubChem CID 2777824
IUPAC Name 4-(trifluoromethylsulfonyl)benzonitrile
SMILES C1=CC(=CC=C1C#N)S(=O)(=O)C(F)(F)F

Specifications

Melting Point 87°C to 92°C
Quantity 1 g
UN Number UN3439
Formula Weight 235.18
Percent Purity ≥97%
Chemical Name or Material 4-(Trifluoromethylsulfonyl)benzonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only