missing translation for 'onlineSavingsMsg'
Learn More

4-Trifluoromethylphenylboronic acid, 98%, Thermo Scientific Chemicals

$66.97 - $66.97

Chemical Identifiers

CAS 128796-39-4
Molecular Formula C7H6BF3O2
Molecular Weight (g/mol) 189.93
MDL Number MFCD00151855
InChI Key ALMFIOZYDASRRC-UHFFFAOYSA-N
Synonym 4-trifluoromethyl phenylboronic acid, 4-trifluoromethylphenylboronic acid, 4-trifluoromethyl benzeneboronic acid, 4-trifluoromethyl phenyl boronic acid, 4-trifluoromethylphenyl boronic acid, 4-trifluoromethyl phenyl boranediol, 4-boronobenzotrifluoride, p-trifluoromethyl phenylboronic acid, 4-trifluoromethylphenyl-boronic acid
PubChem CID 2734389
IUPAC Name [4-(trifluoromethyl)phenyl]boronic acid
SMILES OB(O)C1=CC=C(C=C1)C(F)(F)F
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC359765000
SDS View Documents
Thermo Scientific Chemicals
359765000
500 mg Glass bottle
Each for $66.97
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

128796-39-4
189.93
ALMFIOZYDASRRC-UHFFFAOYSA-N
2734389
OB(O)C1=CC=C(C=C1)C(F)(F)F
C7H6BF3O2
MFCD00151855
4-trifluoromethyl phenylboronic acid, 4-trifluoromethylphenylboronic acid, 4-trifluoromethyl benzeneboronic acid, 4-trifluoromethyl phenyl boronic acid, 4-trifluoromethylphenyl boronic acid, 4-trifluoromethyl phenyl boranediol, 4-boronobenzotrifluoride, p-trifluoromethyl phenylboronic acid, 4-trifluoromethylphenyl-boronic acid
[4-(trifluoromethyl)phenyl]boronic acid

Specifications

128796-39-4
100.0
White
97.5% min. (HPLC)
C7H6BF3O2
500 mg
ALMFIOZYDASRRC-UHFFFAOYSA-N
[4-(trifluoromethyl)phenyl]boronic acid
2734389
98%
4-Trifluoromethylphenylboronic acid
97.5
245.0°C to 250.0°C
Authentic
Glass bottle
MFCD00151855
4-trifluoromethyl phenylboronic acid, 4-trifluoromethylphenylboronic acid, 4-trifluoromethyl benzeneboronic acid, 4-trifluoromethyl phenyl boronic acid, 4-trifluoromethylphenyl boronic acid, 4-trifluoromethyl phenyl boranediol, 4-boronobenzotrifluoride, p-trifluoromethyl phenylboronic acid, 4-trifluoromethylphenyl-boronic acid
OB(O)C1=CC=C(C=C1)C(F)(F)F
189.93
189.93
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only