missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)styrene (stabilized with TBC) 98.0+%, TCI America™

Catalog No. p-7137713
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T33551G 1 g
T33555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T33551G Supplier TCI America Supplier No. T33551G
Only null left

Chemical Identifiers

CAS 402-50-6
Molecular Formula C9H7F3
Molecular Weight (g/mol) 172.15
MDL Number MFCD00075539
InChI Key CEWDRCQPGANDRS-UHFFFAOYSA-N
Synonym 1-trifluoromethyl-4-vinylbenzene, 4-trifluoromethyl styrene, 4-trifluoromethylstyrene, 1-ethenyl-4-trifluoromethyl benzene, 4-vinylbenzotrifluoride, 1-trifluoromethyl-4-vinyl-benzene, 4-trifluoromethystyrene, p-trifluoromethylstyrene, pubchem15485, acmc-1aivr
PubChem CID 2782798
IUPAC Name 1-ethenyl-4-(trifluoromethyl)benzene
SMILES C=CC1=CC=C(C=C1)C(F)(F)F

Specifications

Color Yellow
Boiling Point 66°C
Quantity 1 g
UN Number 1993
Formula Weight 172.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-(Trifluoromethyl)styrene (stabilized with TBC)
TSCA No
Recommended Storage Refrigerator