missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Trifluoromethyl)salicylic Acid 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 178°C |
| Color | White |
| Quantity | 5 g |
| Formula Weight | 206.12 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-(Trifluoromethyl)salicylic Acid |
Safety and Handling
| TSCA | No |