missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)piperidine 97.0+%, TCI America™

$184.82 - $556.38

Chemical Identifiers

CAS 657-36-3
Molecular Formula C6H10F3N
Molecular Weight (g/mol) 153.15
MDL Number MFCD00102068
InChI Key RDRQUUWCJTYHCT-UHFFFAOYSA-N
Synonym 4-trifluoromethyl piperidine, 4-trifluoromethyl-piperidine, piperidine, 4-trifluoromethyl, pubchem22645, 4-trifluormethyl-piperidine
PubChem CID 136469
IUPAC Name 4-(trifluoromethyl)piperidine
SMILES FC(F)(F)C1CCNCC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
T30811G
View Documents
TCI America
T30811G
1 g N/A
Only null left
 
T30815G
View Documents
TCI America
T30815G
5 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

657-36-3
153.15
RDRQUUWCJTYHCT-UHFFFAOYSA-N
136469
FC(F)(F)C1CCNCC1
C6H10F3N
MFCD00102068
4-trifluoromethyl piperidine, 4-trifluoromethyl-piperidine, piperidine, 4-trifluoromethyl, pubchem22645, 4-trifluormethyl-piperidine
4-(trifluoromethyl)piperidine

Specifications

657-36-3
135°C
MFCD00102068
1993
RDRQUUWCJTYHCT-UHFFFAOYSA-N
4-(trifluoromethyl)piperidine
136469
≥97.0% (GC,T)
4-(Trifluoromethyl)piperidine
Yellow
C6H10F3N
1 g
4-trifluoromethyl piperidine, 4-trifluoromethyl-piperidine, piperidine, 4-trifluoromethyl, pubchem22645, 4-trifluormethyl-piperidine
FC(F)(F)C1CCNCC1
153.15
153.15
Liquid
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'