accessibility menu, dialog, popup

UserName

FFFNH

CAS RN 657-36-3

IUPAC Name: 4-(trifluoromethyl)piperidine
Synonyms: 4-(trifluoromethyl)piperidine
Molecular Weight (g/mol): 153.15
Molecular Formula: C6H10F3N
InChi Key: RDRQUUWCJTYHCT-UHFFFAOYSA-N
SMILES: FC(F)(F)C1CCNCC1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (5)
  • (2)
  • (1)
  • (2)
  • (2)